Learn More
C.L. Sturkey Disposable Microtome Blade Dispenser
Catalog No. 22210060 Shop All CL Sturkey Products
Change view
Click to view available options
Product Type:
High Profile Dispenser
Low Profile Dispenser
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-210-060 Low Profile Dispenser
22-210-059 High Profile Dispenser
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-210-060 Supplier C.L. Sturkey Supplier No. DT315D50
Add to Cart
Add to Cart
For dispensing Diamond Microtome blades
  • Used for dispensing Diamond Microtome blades
  • For disposable and permanent microtome knives
Specifications
Product Type Low Profile Dispenser
For Use With (Equipment) Diamond Microtome Blade
For Use With (Application) High and low profile dispensers for disposable and permanent microtome knives