Learn More
C.L. Sturkey Gold Disposable Microtome Blades
Catalog No. 22210046 Shop All CL Sturkey Products
Change view
Click to view available options
Profile:
High
Low
Quantity:
100 Pk.
170 Pk.
50 Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Profile Quantity
22-210-046 High 50 Pk.
22-210013 High 100 Pk.
22-210009 Low 170 Pk.
22-210-049 Low 50 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-210-046 Supplier C.L. Sturkey Supplier No. D554G50
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
The ultimate blade for cutting hard tissue and bone
High-performance specialty blade, tough and durable
Packaging Options
  • Standard Dispenser: Holds 50 blades per dispenser
  • Bulk Dispenser: Holds 120 Heavy Duty High-Profile or 200 Low-Profile blades per dispenser
  • Box Packaging: Holds 100 Heavy Duty High-Profile or 170 Low-Profile blades per box (non-dispensing)
Specifications
Profile High
Disposable Single-use
Quantity 50 Pk.
Coating Titanium Nitride