Learn More
C.L. Sturkey Monarch Microtome Blades
Catalog No. 22210100 Shop All CL Sturkey Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
22-210-100 Stainless Steel
1 options
This item is not returnable. View return policy
Catalog No. 22-210-100 Supplier C.L. Sturkey Supplier No. DT315M50
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
For general paraffin and cryostat use
  • Finely honed stainless steel blades
  • Ground for durability and hardness at a 35 degree angle
  • PTFE overcoating; a special Teflon-like coating that provides a perfectly smooth and slippery surface for ultimate performance
  • Disposable
  • Low profile
  • Numberf blades: 50/Pk.
All general paraffin cutting including: soft tissue, hard tissue, decalcified bone, small biopsies and large tissue
Specifications
Description Low Surface Finish
Product Type Microtome Blade
For Use With (Application) All General Paraffin Cutting including Soft Tissue, Hard Tissue, Decalcified Bone, Small Biopsies and Large Tissue
Material Stainless Steel
Disposable Single-use
Dimensions (L x W x H) 76 x 8 x 0.25 mm