Learn More
C.L. Sturkey Select Disposable Microtome Blades
GSA_VA
Catalog No. 22210052 Shop All CL Sturkey Products
Change view
Click to view available options
Profile:
High
Low
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Profile
22-210-052 High
22-210-051 Low
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-210-052 Supplier C.L. Sturkey Supplier No. DT554S50
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ideal for all general paraffin applications and cryostat use
  • State-of-the-art grinding and coating techniques are specially designed to create an edge that produces superior sections with every cut
  • Standard Dispenser holds 50 blades per pack
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Profile High
For Use With (Equipment) Microtome Blade
Disposable Single-use
Quantity 50 Pk.