Learn More
C.L. Sturkey Silver Disposable Microtome Blades
GSA_VA
Catalog No. 22210047 Shop All CL Sturkey Products
Change view
Click to view available options
Profile:
High
Low
Quantity:
170 Pk.
50 Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Profile Quantity
22-210-047 High 50 Pk.
22-210008 Low 170 Pk.
22-210-050 Low 50 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-210-047 Supplier C.L. Sturkey Supplier No. D554R50
Add to Cart
Add to Cart
This item is not returnable. View return policy
Durable and long-lasting, ideal for general paraffin and cryostat use
Coated with a nitride and copolymer finish to provide smooth, wrinkle-free sections
Packaging Options
  • Standard Dispenser: Holds 50 blades per dispenser
  • Bulk Dispenser: Holds 200 Low-Profile blades per dispenser
  • Box Packaging: Holds 170 Low-Profile blades per box (non-dispensing)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Profile High
Disposable Single-use
Quantity 50 Pk.
Coating Nitride and Copolymer