Learn More
BIOHIPPO c-LEcta DENARASE®ELISA kit
SDP
Supplier:  BIOHIPPO BHE165000021
Encompass
With the DENARASE® ELISA Kit you can detect and quantify up to 30 times lower endonuclease concentrations compared to competitor ELISA Kits. This makes the assay extremely reliable, even when expected concentrations are at a very low level.
Catalog No. NC2737240