Learn More
Enterprise Technology Solutions C-Line Name Badge Holder Kit, Top Load, 3"x4", 20/BX, Clear
SDP
Catalog No. 5000833107
Encompass_Preferred
Magnetic Name Badge Holder Kit includes magnet-style badge holders that secure name badge to your clothing without pins or clips. Magnet attaches easily and won't mar clothing. Complete with reusable clear vinyl holders, inserts and instructions. Top-loading badge holders are sealed on three sides. Inserts are compatible with inkjet and laser printers.
Catalog No. 50-008-33107 Supplier Enterprise Technology Solutions Supplier No. CLI92943
Magnetic Name Badge Holder Kit includes magnet-style badge holders that secure name badge to your clothing without pins or clips. Magnet attaches easily and won't mar clothing. Complete with reusable clear vinyl holders, inserts and instructions. Top-loading badge holders are sealed on three sides. Inserts are compatible with inkjet and laser printers.