Learn More
Enterprise Technology Solutions C-Line Power Extension/Cord Cover, 16 Gauge, 10 Ft, Black
SDP
Catalog No. 5000740672
Encompass_Preferred
An extension cord and cord cover are combined into one convenient unit to save space in office and industrial environments. Durable to withstand heavy foot traffic, this heavy-duty, 16-gauge cord (rated at 13 amp, 125 volt) is covered by flexiduct PVC, a moisture-resistant material that is thermally fused. Unit is ready to use with its grounded, AC duplex receptacle. A lead cord extends 2' beyond the cord cover. UL listed unit is rated at 13 amp, 125 volt.
Catalog No. 50-007-40672 Supplier Enterprise Technology Solutions Supplier No. CLI79101
Add to Cart
Add to Cart
An extension cord and cord cover are combined into one convenient unit to save space in office and industrial environments. Durable to withstand heavy foot traffic, this heavy-duty, 16-gauge cord (rated at 13 amp, 125 volt) is covered by flexiduct PVC, a moisture-resistant material that is thermally fused. Unit is ready to use with its grounded, AC duplex receptacle. A lead cord extends 2' beyond the cord cover. UL listed unit is rated at 13 amp, 125 volt.