Learn More
Enterprise Technology Solutions C-Line PVC ID Badge Cards, 3-3/8"X2-1/8", 100/PK, White
SDP
Catalog No. 5000832610
Encompass_Preferred
Video-grade PVC cards are perfect for creating ID badges in-house with thermal dye-sublimation printers. Each CR80 size card offers a 30 mil thickness. Cards are compatible with leading ID printers including Frago, Zebra, Magicard and Nisca.
Catalog No. 50-008-32610 Supplier Enterprise Technology Solutions Supplier No. CLI89007
Video-grade PVC cards are perfect for creating ID badges in-house with thermal dye-sublimation printers. Each CR80 size card offers a 30 mil thickness. Cards are compatible with leading ID printers including Frago, Zebra, Magicard and Nisca.