Learn More
Enterprise Technology Solutions C-Line Sheet Protectors, Standard, 8-1/2"x11", 100/BX, Clear
SDP
Catalog No. 5000832656
Encompass_Preferred
Sheet protectors offer a top-loading design, being sealed on three sides. No punching of sheets is necessary. Load the sheet while the protector remains in the binder. Sheet protectors fit standard three-ring binders. Polypropylene material offers safe, archival quality, no photocopy-transfer protection. Strong, reinforced binding edge prevents tears with heavy use.
Catalog No. 50-008-32656 Supplier Enterprise Technology Solutions Supplier No. CLI62027
Sheet protectors offer a top-loading design, being sealed on three sides. No punching of sheets is necessary. Load the sheet while the protector remains in the binder. Sheet protectors fit standard three-ring binders. Polypropylene material offers safe, archival quality, no photocopy-transfer protection. Strong, reinforced binding edge prevents tears with heavy use.