Learn More
Medchemexpress LLC c-Met-IN-15 | 330572-32-2 | 98.5% | 299.26 | 5 MG
SDP
Catalog No. 5000360556
Encompass_Preferred
c-Met-IN-15 (compound S3) is a c-Met kinase inhibitor. It inhibits c-Met kinase activity.
  • Inhibits c-Met kinase activity of 21.1% at the concentration of 10 μM.
  • For research use only.
Catalog No. 50-003-60556 Supplier Medchemexpress LLC Supplier No. HY1482795MG
c-Met-IN-15 (compound S3) is a c-Met kinase inhibitor. It inhibits c-Met kinase activity.
  • Inhibits c-Met kinase activity of 21.1% at the concentration of 10 μM.
  • For research use only.