Learn More
Motic™ C-mount Camera Adapters
SCI_ED
Catalog No. p-7227368 Shop All Motic Group Products
Change view
Click to view available options
Magnification Power:
0.5X
0.65X
1X
For Use With (Equipment):
BA310MET Series 0.5 in. Chip Sensor
BA310MET Series 2/3 in. Chip Sensor
BA310MET Series, w/o Additional Lens
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power For Use With (Equipment)
01-674-238 0.5X BA310MET Series 0.5 in. Chip Sensor
01-674-239 0.65X BA310MET Series 2/3 in. Chip Sensor
01-674-240 1X BA310MET Series, w/o Additional Lens
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 01-674-238 Supplier Motic™ Supplier No. 1101001901791
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
The C-mount camera adapter allows Moticams to connect with Motic™ trinocular microscopes.
  • Connects your Moticam with your BA310MET Series Microscope to capture imaging
Specifications
Type Camera Adapter
For Use With (Equipment) BA310MET Series 0.5 in. Chip Sensor
Magnification Power 0.5X
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov