Learn More
Motic™ C-mount Photo Eyepiece Adapter
SCI_ED
Catalog No. 01674241 Shop All Motic Group Products
Change view
Click to view available options
Type:
Camera Adapter
Catalog No. Type
01-674-241 Camera Adapter
1 options
This item is not returnable. View return policy
Catalog No. 01-674-241 Supplier Motic™ Supplier No. 1101001901931
Add to Cart
Add to Cart
This item is not returnable. View return policy
A photo eyepiece is required for this adapter to attach a DSLR Camera to a trinocular microscope.
  • Also requires a T-Mount adapter from your camera's manufacturer
Specifications
Type Camera Adapter
For Use With (Application) Attaching DSLR Camera to Microscope
For Use With (Equipment) BA210S/E Series, BA310E/POL/MET Series, BA410E Series, Panthera C2/E2/Cloud/TEC MET/TEC POL Series, PA43 BIO Series, AE31E/2000/2000 MET Series, SMZ171 Series and GM171 Series
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov