Learn More
Santa Cruz Biotechnology C-MYC (9E10) MOUSE MONOCL
Supplier:  Santa Cruz Biotechnology SC40
Encompass
c-Myc (9E10) mouse monoclonal IgG1; Epitope corresponding to amino acids 408-439 within the carboxy terminal domain of c-Myc of human origin; Species: M, R, H, C, D; Applications: WB, IP, IF, IHC(P), FCM; 200ug/mL
Catalog No. NC9639964