Learn More
Medchemexpress LLC C-Myc inhibitor 6 | 2768765-58-6 | 98.0% | 424.30 | 5 MG
SDP
Catalog No. 5000360855
Encompass_Preferred
c-Myc inhibitor 6 (compound A102) is a c-Myc inhibitor. It decreases cancer cell viability and degrades c-Myc protein. It can be used for the research of c-Myc imbalance, such as cancer, cardiovascular diseases, and viral infection.
  • Inhibits cell viability of HL-60 with an IC50 value less than 100 nM
  • Dose-dependently degrades c-Myc protein in HL-60 cells
Catalog No. 50-003-60855 Supplier Medchemexpress LLC Supplier No. HY1488365MG
c-Myc inhibitor 6 (compound A102) is a c-Myc inhibitor. It decreases cancer cell viability and degrades c-Myc protein. It can be used for the research of c-Myc imbalance, such as cancer, cardiovascular diseases, and viral infection.
  • Inhibits cell viability of HL-60 with an IC50 value less than 100 nM
  • Dose-dependently degrades c-Myc protein in HL-60 cells