Learn More
AdipoGen c-Myc mAb rec Fc ms
SDP
Supplier:  AdipoGen AG27B6302C050
Encompass_Preferred
Recombinant Antibody. Recognizes the c-Myc tag. Shows no binding to native c-Myc protein. Applications ELISA, ICC, IHC, WB. Clone TUN219-2C1 Hypermyc – 9E10 improved variant. Isotype Mouse IgG2a. Formulation Liquid. In PBS. The c-Myc or myc-tag is widely used to detect expression of recombinant proteins in bacteria, yeast, insect and mammalian cell systems. Hyper-Myc is an improved engineered anti-c-myc tag antibody. It showed a higher affinity and stability in direct comparison to hybridoma antibody 9E10 anti-c-myc-tag. This antibody can be detected with anti-mouse Fc secondary antibodies.
Catalog No. 50-256-3141