Learn More
Genscript Corporation c-Myc Peptide
Supplier:  Genscript Corporation RP117315
Encompass
c-Myc Peptide is a useful for displacement of c-Myc-tagged fusion proteins bound to anti-c-Myc antibodies in immunoassays. The successful inhibition of antibody binding by c-Myc peptide demonstrates binding is specific. c-Myc Peptide participates in regulation of growth-related gene transcription.
Catalog No. 50-275-117