Learn More
Medchemexpress LLC c-Myc Peptide TFA | 2918768-02-0 | 99.97% | 1317.32 | 5 MG
SDP
Catalog No. 5000424461
Encompass_Preferred
c-Myc Peptide TFA is a synthetic peptide that corresponds to the C-terminal amino acids (410-419) of human c-myc protein. It is involved in the regulation of growth-related gene transcription.
  • Synthetic peptide corresponding to C-terminal amino acids (410-419) of human c-myc protein.
  • Regulates growth-related gene transcription.
  • Plays a critical role in the growth of breast cancer cells in vitro.
Catalog No. 50-004-24461 Supplier Medchemexpress LLC Supplier No. HYP03125MG
c-Myc Peptide TFA is a synthetic peptide that corresponds to the C-terminal amino acids (410-419) of human c-myc protein. It is involved in the regulation of growth-related gene transcription.
  • Synthetic peptide corresponding to C-terminal amino acids (410-419) of human c-myc protein.
  • Regulates growth-related gene transcription.
  • Plays a critical role in the growth of breast cancer cells in vitro.