Learn More
Medchemexpress LLC HY-P0312 1mg Medchemexpress, c-Myc Peptide Trifluoroacetate CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP03121MG
Encompass_Preferred
Medchemexpress, HY-P0312 1mg c-Myc Peptide Trifluoroacetate CAS: c-Myc Peptide Trifluoroacetate is a synthetic peptide corresponding to the C-terminal amino acids (410-419) of human c-myc protein, and participates in regulation of growth-related gene transcription. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4113