Learn More
Sigma Aldrich Fine Chemicals Biosciences C myc proto oncogene human
Catalog No. 502904043
Encompass
MYCnbsponcogene is a global transcription factor. The gene is located on human chromosome 8q24.21. MYCnbsponcogene is a cancer promoting gene.
Catalog No. 50-290-4043 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SRP20895UG
MYCnbsponcogene is a global transcription factor. The gene is located on human chromosome 8q24.21. MYCnbsponcogene is a cancer promoting gene.