Learn More
Enzo Life Sciences c-Myc (human) Recombinant monoclonal antibody (9E10) (Human IgG1 kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4650200
Encompass_Preferred
Clone: 9E10. Isotype: IgG1κ. Immunogen: Synthetic peptide corresponding to aa 408-439 from C-terminus of human c-myc. Species reactivity: Human. Specificity: Recognizes epitope sequence EQKLISEEDL, located in aa 410-419 of human c-myc protein. Applications: IF, IHC (FS), IHC (PS), IP, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2992