Learn More
Bio Rad Laboratories C' PCR Plate Sealing Film, adhesive, optical, 100/pk
Catalog No. NC1818182
Encompass
Use Microseal 'C' Optical Adhesive Sealing Films for all thermal cycling, including real-time PCR, including plates with raised rims. Pressure-sensitive adhesive film sticks to the plate, not your gloves.
Catalog No. NC1818182 Supplier Bio Rad Laboratories Supplier No. MSC1001
Use Microseal 'C' Optical Adhesive Sealing Films for all thermal cycling, including real-time PCR, including plates with raised rims. Pressure-sensitive adhesive film sticks to the plate, not your gloves.