Learn More
Alpco Ltd C-Peptide Chemiluminescence ELISA Lab Pack (5 Plates)
SDP
Supplier:  Alpco Ltd 80CPTHUCH05
Encompass
The Chemi Human C-peptide ELISA Lab Pack is an FDA Registered, In Vitro Diagnostic Use tool for the quantification of human C-peptide in serum and plasma samples in both clinical and research laboratory settings. The ELISA features a broad dynamic range and superior sensitivity and is now offered in a convenient 5 plate package. *Lot dependent as standards are lyophilized. FDA Class I Device. For In Vitro Diagnostic Use.
Catalog No. NC2271690