Learn More
Alpco Ltd C-Peptide ELISA, FDA Registered
SDP
Supplier:  Alpco Ltd 80CPTHUE01.1
Encompass_Preferred
The C-peptide ELISA is an FDA Registered For In Vitro Diagnostic Use tool for the quantification of human C-peptide in serum and plasma samples in both clinical and research laboratory settings.Superior sensitivity of 2.95 pmol/LBroad dynamic range of 20-3,000 pmol/L
Catalog No. 50-922-577