Learn More
Drg International Inc DRG C-Peptide ELISA, 96 Well Kit
Supplier:  Drg International Inc EIA1293
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of c-peptide in serum, plasma (EDTA-, heparin- or citrate plasma), and urine. C-Peptide measurement has several advantages over immunoassays of insulin. The half-life of C-Peptide in the circulation is between two and five times longer than that of insulin. Therefore, C-Peptide levels are a more stable indicator of insulin secretion than the more rapidly changing levels of insulin. The clinical indications for C-Peptide measurement include diagnosis of insulinoma and differentation from factitious hypoglycemia, follow-up of pancreatectomy, and evaluation of viability of islet cell transplants. Recently, these indications have been dramatically expanded to permit evaluation of insulin dependence in maturity onset diabetes mellitus. The DRG C-Peptide ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding.
Catalog No. 50-187-1249