Learn More
Crystal Chem Inc C-Peptide ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80954
Encompass
The C-Peptide ELISA is a highly sensitive assay that is used to quantify levels of human c-peptide in serum and plasma. Highlights: Kit uses sample size of only 25 µL. High sensitivity. Dynamic range of 0.37 - 15 ng/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7860