Learn More
Bachem C-Peptide (human) Trifluoroacetate
SDP
Supplier:  Bachem 4009494.0500
Encompass_Preferred
C-Peptide (human) Trifluoroacetate, H-Glu-Ala-Glu-Asp-Leu-Gln-Val-Gly-Gln-Val-Glu-Leu-Gly-Gly-Gly-Pro-Gly-Ala-Gly-Ser-Leu-Gln-Pro-Leu-Ala-Leu-Glu-Gly-Ser-Leu-Gln-OH, 33017-11-7, C129H211N35O48, Mr 3020.30, 0.5mg. Insulin Precursor (57-87) (human). The measurement of the C-peptide under standardized conditions provides a sensitive, well accepted, and clinically validated assessment of b-cell function. The C-peptide measurement is the most suitable primary outcome for clinical trials of therapies aimed at preserving or improving endogenous insulin secretion in type 1 diabetes patients. C-peptide also shows hormonal activity. Clinical studies demonstrated that C-peptide administration in type 1 diabetes patients, who lack the peptide, alleviates the diabetes-induced renal and nerve dysfunctions.
Catalog No. NC1389244