Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human C-Peptide ELISA Kit

Catalog No. BMS21912
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BMS21912 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BMS21912 Supplier Invitrogen™ Supplier No. BMS21912
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human c-Peptide ELISA quantitates Hu c-Peptide in human serum, plasma, urine, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu c-Peptide. Principle of the method The Human c-Peptide solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, binding to the target on a different epitope from the capture antibody. A conjugated enzyme has been incorporated into the assay. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

After removal of the precursor signal peptide, proinsulin is post-translationally cleaved into three peptides: the B chain and A chain peptides, which are covalently linked via two disulfide bonds to form insulin, and C-peptide. Binding of insulin to the insulin receptor (INSR) stimulates glucose uptake. A multitude of mutant alleles with phenotypic effects have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01308
Assay Range 23-1500 pg/mL
Assay Sensitivity 2.9 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Growth Factors & Receptors
Sample Type Plasma, Serum, Supernatant, Urine
Substrate Type Adaptor Proteins
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Cpeptide; IDDM; IDDM1; IDDM2; ILPR; INS; Ins1; Ins-1; Ins2-rs1; insulin; insulin 1; Insulin A chain; Insulin B chain; insulin I; insulin-1; Insulin-1 A chain; Insulin-1 B chain; IRDN; MODY10; preproinsulin; proinsulinCpeptide; IDDM; IDDM1; IDDM2; ILPR; INS; Ins1; Ins-1; Ins2-rs1; insulin; insulin 1; Insulin A chain; Insulin B chain; insulin I; insulin-1; Insulin-1 A chain; Insulin-1 B chain; IRDN; MODY10; preproinsulin; proinsulin; IDDM; IDDM1; IDDM2; ILPR; INS; IRDN; MODY10
Gene ID (Entrez) 3630
Gene Symbol INS
Interassay CV <10%
Intraassay CV <6%
Kit Contents Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases Connecting Peptide, Insulin, Insulin A chain, Insulin B chain, preproinsulin, proinsulin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL, Urine 50 μL
Shipping Condition Wet Ice
Storage Requirements 2-8°C
Target C-Peptide
Test Time 1 hr 20 min
Total Assay Time 4 hr
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.