Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedical™ C-Peptide IRMA Kit

Catalog No. MP07RK84CT Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP07RK84CT 100 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP07RK84CT Supplier MP Biomedical™ Supplier No. 07RK84CT
Add to Cart
Add to Cart

Quantitative Immunoradiometric assay

The C-Peptide IRMA Kit is intended for the measurement of C-Peptide in a sample. FDA Exempt for In Vitro Diagnostic (IVD) use in the United States. CE Marked for IVD use outside the United States.
TRUSTED_SUSTAINABILITY

Specifications

Description Intended for the measurement of C-Peptide in a sample.
Product Type Kit and Assay
Sample Type Serum
For Use With (Application) Endocrinology
Quantity 100 Tubes
Regulatory Status FDA Exempt
Sample Volume 50 μL
Storage Requirements 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.