Learn More
Alpco Ltd Mouse C-peptide ELISA
SDP
Supplier:  Alpco Ltd 80CPTMSE01
Encompass_Preferred
The Mouse C-peptide ELISA quantifies the C-peptide protein products of mouse I and mouse II proinsulin genes. The assay utilizes a dual-monoclonal antibody sandwich format, bi-level control set, and 96-well microplate comprising removable strips. A single Mouse C-peptide ELISA kit has the performance characteristics and flexibility necessary to confidently measure up to 40 samples in duplicate.
Catalog No. NC1072456