Learn More
Alpco Ltd Rat C-peptide ELISA
SDP
Catalog No. 50922579
Encompass_Preferred
The Rat C-peptide ELISA quantifies the C-peptide protein products of rat I and rat II proinsulin genes.Only 10 ¿L of sample is sufficient for the assaySuperior sensitivity of 10.8 pMDynamic range of 50-4,500 pM
Catalog No. 50-922-579 Supplier Alpco Ltd Supplier No. 80CPTRTE01
The Rat C-peptide ELISA quantifies the C-peptide protein products of rat I and rat II proinsulin genes.Only 10 ¿L of sample is sufficient for the assaySuperior sensitivity of 10.8 pMDynamic range of 50-4,500 pM