Learn More
Sino Biological Anti-C. Perfringens Neuraminidase Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11680MM0420
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Clostridium Perfringens Neuraminidase (Catalog#11680-V07E; P10481.1; Cys 2-Gln 382). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4627