Learn More
Sino Biological Recombinant Anti-C. Perfringens Neuraminidase Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11680R11220
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant C. perfringens Clostridium Perfringens Neuraminidase (Catalog#11680-V07E; P10481.1; Cys2-Gln 382).
Catalog No. 50-255-4625