Learn More
Sino Biological Anti-C. Perfringens Neuraminidase Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11680RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Clostridium Perfringens Neuraminidase (rClostridium Perfringens Neuraminidase; Catalog#11680-V07E; P10481.1; Cys 2-Gln 382). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4624