Learn More
Enzo Life Sciences AEC peroxidase substrate kit (300ml)
SDP
Catalog No. 502012559
Encompass_Preferred
The AEC Peroxidase Substrate Kit contains reagents for visualization of peroxidase reactions using Aminoethylcarbazole (AEC). In the presence of H2O2, peroxidases catalyze the oxidation of AEC into a brick red precipitate. The supplied reagents are sufficient for preparation of 300 ml of ready-to-use color reaction mixture. Applications: Colorimetric detection .
Catalog No. 50-201-2559 Supplier Enzo Life Sciences Supplier No. ENZ43825
The AEC Peroxidase Substrate Kit contains reagents for visualization of peroxidase reactions using Aminoethylcarbazole (AEC). In the presence of H2O2, peroxidases catalyze the oxidation of AEC into a brick red precipitate. The supplied reagents are sufficient for preparation of 300 ml of ready-to-use color reaction mixture. Applications: Colorimetric detection .