Learn More
Medchemexpress LLC C-Phycocyanin | 11016-15-2 | 70,000-110,000 | 100 MG
SDP
Catalog No. 5000428459
Encompass_Preferred
C-Phycocyanin (C-PC) is a protein pigment extensively used as a nutrient supplement for human beings and for diverse research applications. It is supplied as a solid, appearing light blue to blue, and is shipped at room temperature.
  • Used as a nutrient supplement.
  • Acts as a protein pigment.
  • Supports various research applications, including in vitro and in vivo studies.
  • Can decrease CDK-4 mRNA levels.
  • Maximizes caspase-3 expression.
  • Has shown to decrease tumor weights in studies.
  • Increases spleen weight and promotes immune organ growth.
  • Enhances immune function.
  • Inhibits Cyclin D1 expression.
  • Exhibits synergistic effects when combined with All-trans retinoic acid (ATRA).
Catalog No. 50-004-28459 Supplier Medchemexpress LLC Supplier No. HYD1025100MG
C-Phycocyanin (C-PC) is a protein pigment extensively used as a nutrient supplement for human beings and for diverse research applications. It is supplied as a solid, appearing light blue to blue, and is shipped at room temperature.
  • Used as a nutrient supplement.
  • Acts as a protein pigment.
  • Supports various research applications, including in vitro and in vivo studies.
  • Can decrease CDK-4 mRNA levels.
  • Maximizes caspase-3 expression.
  • Has shown to decrease tumor weights in studies.
  • Increases spleen weight and promotes immune organ growth.
  • Enhances immune function.
  • Inhibits Cyclin D1 expression.
  • Exhibits synergistic effects when combined with All-trans retinoic acid (ATRA).