Learn More
Athens Research And Technology Inc C-Reactive Protein (CRP), Native Human Plasma, ≥95% (SDS-PAGE), 1 mg
Catalog No. NC3244544
Encompass_Preferred
C-Reactive Protein (CRP) is a native protein purified from human plasma with ≥95% (SDS-PAGE), supplied frozen in a 1 mg pack. Molecular weight: 24 kDa.
Catalog No. NC3244544 Supplier Athens Research And Technology Inc Supplier No. 16160318161MG
C-Reactive Protein (CRP) is a native protein purified from human plasma with ≥95% (SDS-PAGE), supplied frozen in a 1 mg pack. Molecular weight: 24 kDa.