Learn More
CPC Scientific H-Lys-Pro-Gln-Leu-Trp-Pro-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific CRP003A
Encompass
An internally quenched peptide substrate for calpain-1 with optimal cleavage motifs flanking the scissile bond, interferes with the membrane-associated oxidative metabolism in human neutrophilsSEQUENCE: H-Lys-Pro-Gln-Leu-Trp-Pro-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: KPQLWPMOLECULAR FORMULA: C38H57N9O8MOLECULAR WEIGHT: 767.93STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [130348-99-1]SYNONYMS: RESEARCH AREA: CancerREFERENCES:R.M.Heuertz et al., J. Immunol., 156, 3412 (1996)SKU(s): CRP-003A, CRP-003B
Catalog No. 50-210-9175