Learn More
CPC Scientific H-Val-Gly-Gly-Ser-Glu-Ile-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific CRP001A
Encompass
The peptide interferes with the membrane-associated oxidative metabolism in human neutrophils.SEQUENCE: H-Val-Gly-Gly-Ser-Glu-Ile-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: VGGSEIMOLECULAR FORMULA: C23H40N6O10MOLECULAR WEIGHT: 560.61STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [130349-01-8]SYNONYMS: RESEARCH AREA: CancerREFERENCES:R.M.Heuertz et al., J. Immunol., 156, 3412 (1996)SKU(s): CRP-001A, CRP-001B
Catalog No. 50-210-9171