Learn More
Drg International Inc C-Reactive Protein Hs Elisa
Supplier:  Drg International Inc EIA3954
Encompass_Preferred
A high sensitivity enzyme immunoassay for the quantitative determination of C-reactive protein (CRP) concentration in serum. CRP is synthesized in the liver and is normally present as a trace constituent of serum or plasma at levels less than 0.3 mg/dL. CRP is one of the acute-phase proteins, the serum or plasma levels of which rise during general, nonspecific response to a wide variety of diseases. This include infections by gram-positive and gram-negative organisms, acute phase of rheumatoid arthritis, abdominal abscesses, and inflammation of the bile duct. Additionally, measurement of CRP by high-sensitivity CRP assays may add to the predictive value of other cardiac markers (myoglobin, creatine-kinase-MB, troponin I and T), which are used to assess the risk of cardiovascular and peripheral vascular disease. The hsCRP ELISA is based on the principle of a solid phase enzyme-linked immunosorbent assay.
Catalog No. 50-194-7602