Learn More
Cayman Chemical C-ReactIve ProteInhumnre 200ug
SDP
Catalog No. 502919314
Encompass_Preferred
Source: Recombinant human C-reactive protein expressed in HEK293 cells • Amino acids: 19-224 • MW: 23 kDa
Catalog No. 50-291-9314 Supplier Cayman Chemical Supplier No. 37018200 UG
Source: Recombinant human C-reactive protein expressed in HEK293 cells • Amino acids: 19-224 • MW: 23 kDa