Learn More
Carbosynth LLC. Cyclo[Arg-Gly-Asp-D-Phe-Lys(Biotin)], 1mg, MW: (M.W. 829.98), Molecular formula: C37H55N11O9S
SDP
Supplier:  Carbosynth LLC. PCI3895PI1MG
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Synonyms: cyclo [Arg-Gly-Asp-D-Phe-Lys(Biotin)], c[RGDfK(Biotin)], Application: RGD Peptide equipped with a biotin reporting tag, Storage Temperature: -20C, References: D. Boturyn, J.-L. Coll, E. Garager, M.-C. Favrot, and P. Dumy, JACS, 126, 5730 (2004).
Catalog No. 50-168-6313