Learn More
Carbosynth LLC. Cyclo[Arg-Gly-Asp-D-Phe-Lys(Biotin-PEG-PEG)], 5mg, MW: (M.W. 1120.30), Molecular formula: C49H77N13O15S
SDP
Supplier:  Carbosynth LLC. PCI3697PI5MG
Encompass_Preferred
Synonyms: cyclo [Arg-Gly-Asp-D-Phe-Lys(Biotin-PEG-PEG)], c(RGDfK(Biotin-PEG-PEG)) where PEG = 8-Amino-3,6-Dioxaoctanoic Acid, cRGDfK-(Biotin-PEG-PEG), Application: RGD Peptide equipped with a biotin reporting tag and PEG spacers for more efficient binding to lipid surfaces (Requires further derivatization before use), Storage Temperature: -20°C, References: D. Boturyn, J.-L. Coll, E. Garager, M.-C. Favrot, and P. Dumy, JACS, 126, 5730 (2004)., C. Dolce, A. Vakani, L. Archer, J.A. Morris-Wiman, and L.S. Holliday, J. Dent. Res., 82, 682 (2003)., P. Holig, M. Bach, T. Volkel, T. Nahde, S. Hoffmann, R. Muller, and R.E. Kontermann, Protein Engin. Design Selection, 17, 433 (2004).
Catalog No. 50-168-6292