Learn More
Carbosynth LLC. Cyclo[Arg-Gly-Asp-D-Phe-Lys(Cys)], 1mg, MW: (M.W. 706.83), Molecular formula: C30H46N10O8S
SDP
Supplier:  Carbosynth LLC. RGD3794PI1MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: cyclo [Arg-Gly-Asp-D-Phe-Lys(Cys)], c[RGDfK(C)], Application: RGD Peptide for Radiolabeling and Imaging, Storage Temperature: -20C, References: E. Morales-Avila, et al., Bioconjugate Chem., 22, 913 (2011)., A. Vilchis-Juárez, et al., J. of Biomedical Nanotechnology, 10, 393 (2014).
Catalog No. 50-168-6947