Learn More
Vici Precision Sampling, Inc. C SERIES GAS SYRINGE 1ML
Catalog No. NC9787613
Encompass_Preferred
Gas Syringe C 1ml RN .029 in. x .012 in. x 2 in. bevel, open end
Catalog No. NC9787613 Supplier Vici Precision Sampling, Inc. Supplier No. 030033
Gas Syringe C 1ml RN .029 in. x .012 in. x 2 in. bevel, open end