Learn More
Research Products International Corp C Shaped Lead Ring Flask Weight, Vinyl Coated, Open, 1.9 Inch Inner Diameter for 125-500ml Flasks, 1/2 Pound
SDP
Catalog No. 50550318
Encompass
C Shaped Lead Ring helps stabilize glass and plastic containers. Rings fit around neck of flasks and eliminate floating in water baths or tipping on benchtops. Open ring design allows adjustment to fit wide vessels. Vikem vinyl coating protects glassware and is resistant to most acids and solvents.
WARNING  This product can expose you to chemicals including Lead, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-550-318 Supplier Research Products International Corp Supplier No. 248856
C Shaped Lead Ring helps stabilize glass and plastic containers. Rings fit around neck of flasks and eliminate floating in water baths or tipping on benchtops. Open ring design allows adjustment to fit wide vessels. Vikem vinyl coating protects glassware and is resistant to most acids and solvents.
WARNING  This product can expose you to chemicals including Lead, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.