Learn More
Nanoentek Inc C-Slides Cell counting chamber slides, Pack of 50
SDP
Catalog No. NC2583275
Encompass
No need for coverslips. - No need for washing (Disposable) - Reduce exposure to infectious samples and contamination - Reproducibility and reliability guaranteed - Made of sturdy quartz grade optical plastic
Catalog No. NC2583275 Supplier Nanoentek Inc Supplier No. CS050
No need for coverslips. - No need for washing (Disposable) - Reduce exposure to infectious samples and contamination - Reproducibility and reliability guaranteed - Made of sturdy quartz grade optical plastic