Learn More
Santa Cruz Biotechnology Anti-c-Src/Src Antibody (B-12), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC8056
Encompass
Anti-c-Src/Src Antibody (B-12) is recommended for detection of c-Src, Yes p62, Fyn p59, c-Fgr p55 and c-Src-2 of mouse, rat and human origin by WB, IP, IF, IHC(P) and ELISA.
Catalog No. NC0944918