Learn More
ECM Bioscience c-Src Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Supplier:  ECM Bioscience SK6350
Encompass_Preferred
The c-Src phospho-regulation antibody sampler kit can be used to detect c-Src phosphorylation on Tyr-215 and Tyr-530. The kit also includes a monoclonal antibody to monitor total c-Src expression levels and secondary reagents for detection of these antibodies. Applications: WB, E; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8835