Learn More
Ace Glass, Inc. PTFE stir blade, 24x130mm, 10mm shafts
SDP
Catalog No. 509558249
Encompass_Preferred
24x130mm PTFE stir blade for 10mm O.D. button type stir shafts.
Catalog No. 50-955-8249 Supplier Ace Glass, Inc. Supplier No. 808519
24x130mm PTFE stir blade for 10mm O.D. button type stir shafts.